Learn More
Grainger Electric Air Compressor 10 hp 2 Stage
Catalog No. 502766356
Encompass_Preferred
The Ingersoll Rand 10 HP Electric Air Compressor is a single-stage, oil-lubricated, belt-driven air compressor. It has a 10 horsepower electric motor and delivers 16.5 CFM at 175 PSI with a 20 gallon vertical ASME air receiver tank. This compressor is designed for applications requiring high pressures like automotive service, body shops, machine shops, production lines, and other manufacturing. It features a cast iron, V-shaped cylinder for efficient cooling. The pump and motor are mounted on a heavy-duty steel base to reduce vibration. This air compressor requires a 230 volt 1 phase 60 hertz electrical connection and includes an automatic tank drain valve. The model number is 2475N7.5.
Catalog No. 50-276-6356 Supplier Grainger Supplier No. 16V895
Add to Cart
Add to Cart
Request a Quote
The Ingersoll Rand 10 HP Electric Air Compressor is a single-stage, oil-lubricated, belt-driven air compressor. It has a 10 horsepower electric motor and delivers 16.5 CFM at 175 PSI with a 20 gallon vertical ASME air receiver tank. This compressor is designed for applications requiring high pressures like automotive service, body shops, machine shops, production lines, and other manufacturing. It features a cast iron, V-shaped cylinder for efficient cooling. The pump and motor are mounted on a heavy-duty steel base to reduce vibration. This air compressor requires a 230 volt 1 phase 60 hertz electrical connection and includes an automatic tank drain valve. The model number is 2475N7.5.