Learn More
Grainger Electric Air Compressor 10 hp 2 Stage
Catalog No. 502766380
Encompass_Preferred
The Ingersoll Rand 10 HP Electric Air Compressor is a single-stage, oil-lubricated, belt-driven air compressor that provides 125 PSI max pressure with a 16 gallon vertical ASME tank. It has a cast iron cylinder and pump, and is powered by a 10 HP, 230V, 1-phase, 60 Hz electric motor. This compressor can deliver 18.1 CFM at 90 PSI and has an operating pressure range of 80 to 125 PSI. It features an automatic start/stop control and thermal overload protection. This air compressor is designed for applications like automotive service, manufacturing, construction, wood working shops, auto body shops, tire shops, and general maintenance and repair.
Catalog No. 50-276-6380 Supplier Grainger Supplier No. 16V892
Add to Cart
Add to Cart
Request a Quote
The Ingersoll Rand 10 HP Electric Air Compressor is a single-stage, oil-lubricated, belt-driven air compressor that provides 125 PSI max pressure with a 16 gallon vertical ASME tank. It has a cast iron cylinder and pump, and is powered by a 10 HP, 230V, 1-phase, 60 Hz electric motor. This compressor can deliver 18.1 CFM at 90 PSI and has an operating pressure range of 80 to 125 PSI. It features an automatic start/stop control and thermal overload protection. This air compressor is designed for applications like automotive service, manufacturing, construction, wood working shops, auto body shops, tire shops, and general maintenance and repair.