Learn More
Grainger Electric Air Compressor 10 hp 2 Stage
Catalog No. 502766355
Encompass_Preferred
The Ingersoll Rand 10 HP 2-Stage Electric Air Compressor is a powerful and reliable air compressor for industrial use. It features a 10 horsepower motor that delivers 175 max PSI and 21.2 CFM at 175 PSI. This 2-stage compressor has a 60 gallon vertical air receiver tank constructed of ASME certified steel. It is equipped with an integrated dryer to remove moisture. The electric motor requires a 230 volt 3 phase power supply. This industrial grade air compressor is designed for continuous use applications. Its durable construction and high air output make it well suited for automotive service, manufacturing, construction and other demanding environments.
Catalog No. 50-276-6355 Supplier Grainger Supplier No. 16V893
Add to Cart
Add to Cart
Request a Quote
The Ingersoll Rand 10 HP 2-Stage Electric Air Compressor is a powerful and reliable air compressor for industrial use. It features a 10 horsepower motor that delivers 175 max PSI and 21.2 CFM at 175 PSI. This 2-stage compressor has a 60 gallon vertical air receiver tank constructed of ASME certified steel. It is equipped with an integrated dryer to remove moisture. The electric motor requires a 230 volt 3 phase power supply. This industrial grade air compressor is designed for continuous use applications. Its durable construction and high air output make it well suited for automotive service, manufacturing, construction and other demanding environments.