Learn More
Grainger Electric Air Compressor 15 hp 2 Stage
Catalog No. 502767417
Encompass_Preferred
The Ingersoll Rand 15 HP Electric Air Compressor (Model 2475N7.5) is an industrial-grade compressor with a 15 HP motor, max pressure of 175 PSI, and 103 CFM at 90 PSI. It features a two-stage, oil-lubricated system with a 60-gallon ASME certified steel tank. Equipped with an aftercooler and moisture separator, it requires a 230V 3-phase power supply. Measuring 48 x 32 x 75 inches and weighing 990 pounds, it's ideal for auto repair shops or manufacturing facilities needing high capacity.
Catalog No. 50-276-7417 Supplier Grainger Supplier No. 16V897
Add to Cart
Add to Cart
Request a Quote
The Ingersoll Rand 15 HP Electric Air Compressor (Model 2475N7.5) is an industrial-grade compressor with a 15 HP motor, max pressure of 175 PSI, and 103 CFM at 90 PSI. It features a two-stage, oil-lubricated system with a 60-gallon ASME certified steel tank. Equipped with an aftercooler and moisture separator, it requires a 230V 3-phase power supply. Measuring 48 x 32 x 75 inches and weighing 990 pounds, it's ideal for auto repair shops or manufacturing facilities needing high capacity.