Learn More
Grainger Electric Air Compressor 15 hp 2 Stage
Catalog No. 502766170
Encompass_Preferred
The Ingersoll Rand 10 HP Electric Air Compressor is a single-stage, oil-lubricated, belt-driven unit with a 10 horsepower motor. It delivers 16 CFM at 175 PSI and includes a 20-gallon vertical ASME tank. Ideal for high-pressure needs in automotive, body shops, and production lines, it features a cast iron, V-shaped cylinder for cooling and an oil-lubricated pump for reduced wear. The 230-volt motor ensures reliable power, making it suitable for demanding commercial and industrial applications.
Catalog No. 50-276-6170 Supplier Grainger Supplier No. 16V896
Add to Cart
Add to Cart
Request a Quote
The Ingersoll Rand 10 HP Electric Air Compressor is a single-stage, oil-lubricated, belt-driven unit with a 10 horsepower motor. It delivers 16 CFM at 175 PSI and includes a 20-gallon vertical ASME tank. Ideal for high-pressure needs in automotive, body shops, and production lines, it features a cast iron, V-shaped cylinder for cooling and an oil-lubricated pump for reduced wear. The 230-volt motor ensures reliable power, making it suitable for demanding commercial and industrial applications.