Learn More
Grainger Electric Air Compressor 5 hp 2 Stage
Catalog No. 502766172
Encompass_Preferred
The Ingersoll Rand 5 HP 60 Gallon Two-Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade compressor designed for high performance and durability. It features a 5 HP motor with a maximum pressure of 175 PSI and a pump capacity of 15.2 CFM at 90 PSI. The 60-gallon vertical ASME tank is made of durable steel. It has an oil sight glass, oil drain valve, and an automatic tank drain valve. The oil-lubricated pump ensures cooler operation and reduced wear. It requires a 230V 1-phase power supply. Dimensions: 30" L x 34" W x 75" H; weight: 415 lbs.
Catalog No. 50-276-6172 Supplier Grainger Supplier No. 16V888
The Ingersoll Rand 5 HP 60 Gallon Two-Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade compressor designed for high performance and durability. It features a 5 HP motor with a maximum pressure of 175 PSI and a pump capacity of 15.2 CFM at 90 PSI. The 60-gallon vertical ASME tank is made of durable steel. It has an oil sight glass, oil drain valve, and an automatic tank drain valve. The oil-lubricated pump ensures cooler operation and reduced wear. It requires a 230V 1-phase power supply. Dimensions: 30" L x 34" W x 75" H; weight: 415 lbs.