Learn More
Grainger Electric Air Compressor 5 hp 2 Stage
Catalog No. 502766174
Encompass_Preferred
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade air compressor designed for high performance and durability. It features a 7.5 HP electric motor with a two-stage pump that delivers 16.5 CFM at 175 PSI with a maximum pressure of 175 PSI. This 60 gallon vertical tank provides superior air storage. It has a oil-lubricated pump for reduced wear and tear. This compressor includes an electric magnetic starter for motor protection. The pump and motor are mounted on a heavy-duty steel base with vibration isolation feet. This compressor requires a 230V 1-phase power supply.
Catalog No. 50-276-6174 Supplier Grainger Supplier No. 16V899
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade air compressor designed for high performance and durability. It features a 7.5 HP electric motor with a two-stage pump that delivers 16.5 CFM at 175 PSI with a maximum pressure of 175 PSI. This 60 gallon vertical tank provides superior air storage. It has a oil-lubricated pump for reduced wear and tear. This compressor includes an electric magnetic starter for motor protection. The pump and motor are mounted on a heavy-duty steel base with vibration isolation feet. This compressor requires a 230V 1-phase power supply.