Learn More
Grainger Electric Air Compressor 5 hp 2 Stage
Catalog No. 502767047
Encompass_Preferred
The Ingersoll Rand Type 30 Electric Air Compressor is a single-stage, oil-free, reciprocating air compressor. It has a 2 horsepower electric motor and delivers 5.3 CFM at 90 PSI with a maximum pressure of 135 PSI. This compressor has an aluminum cylinder head and a cast iron pump for durability. It is belt driven and has an automatic start/stop control. The tank capacity is 30 gallons. This compressor requires a 230 volt power supply and is designed for applications such as automotive service, wood working shops, and general industry.
Catalog No. 50-276-7047 Supplier Grainger Supplier No. 16V889
The Ingersoll Rand Type 30 Electric Air Compressor is a single-stage, oil-free, reciprocating air compressor. It has a 2 horsepower electric motor and delivers 5.3 CFM at 90 PSI with a maximum pressure of 135 PSI. This compressor has an aluminum cylinder head and a cast iron pump for durability. It is belt driven and has an automatic start/stop control. The tank capacity is 30 gallons. This compressor requires a 230 volt power supply and is designed for applications such as automotive service, wood working shops, and general industry.