Learn More
Grainger Electric Air Compressor 5 hp 2 Stage
Catalog No. 502766360
Encompass_Preferred
The Ingersoll Rand 5 HP 60 Gallon Two-Stage Electric Air Compressor is ideal for industrial use, featuring a 5 HP motor delivering 16.5 CFM at 175 PSI. Its 60-gallon vertical ASME air receiver allows extended run times. The cast iron, two-stage pump with a V-shaped cylinder ensures smooth operation. Safety is enhanced with a fully enclosed belt guard and electric motor. An aftercooler reduces moisture in the air stream. Easy-to-use controls and a pressure gauge monitor tank pressure, making it suitable for demanding industrial applications requiring high airflow.
Catalog No. 50-276-6360 Supplier Grainger Supplier No. 16V898
The Ingersoll Rand 5 HP 60 Gallon Two-Stage Electric Air Compressor is ideal for industrial use, featuring a 5 HP motor delivering 16.5 CFM at 175 PSI. Its 60-gallon vertical ASME air receiver allows extended run times. The cast iron, two-stage pump with a V-shaped cylinder ensures smooth operation. Safety is enhanced with a fully enclosed belt guard and electric motor. An aftercooler reduces moisture in the air stream. Easy-to-use controls and a pressure gauge monitor tank pressure, making it suitable for demanding industrial applications requiring high airflow.