Learn More
Grainger Electric Air Compressor 7.5 hp 2 Stage
Catalog No. 502766390
Encompass_Preferred
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is a powerful, industrial-grade air compressor designed for professional use. It features a 7.5 HP electric motor with a max pressure of 175 PSI and a 60 gallon vertical air receiver tank. This two stage compressor delivers 14.0 CFM at 90 PSI and 17.9 CFM at 40 PSI. It has a cast iron, two cylinder pump with a thermal overload protector. The compressor includes an ASME coded air receiver tank, a pressure gauge, and an automatic tank drain valve. This model weighs approximately 415 lbs and measures 77 x 32 x 75 inches.
Catalog No. 50-276-6390 Supplier Grainger Supplier No. 16V887
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is a powerful, industrial-grade air compressor designed for professional use. It features a 7.5 HP electric motor with a max pressure of 175 PSI and a 60 gallon vertical air receiver tank. This two stage compressor delivers 14.0 CFM at 90 PSI and 17.9 CFM at 40 PSI. It has a cast iron, two cylinder pump with a thermal overload protector. The compressor includes an ASME coded air receiver tank, a pressure gauge, and an automatic tank drain valve. This model weighs approximately 415 lbs and measures 77 x 32 x 75 inches.