Learn More
Grainger Electric Air Compressor 7.5 hp 2 Stage
Catalog No. 502767728
Encompass_Preferred
The Ingersoll Rand Type 30 Reciprocating Air Compressor is a 2 HP electric powered air compressor that provides 7.0 CFM at 90 PSI with a 30 gallon ASME vertical air receiver tank. It has a 115V 1-phase motor with automatic thermal overload protection and pressure switch with gauge. This air compressor is oil-free with a cast iron reciprocating pump and is designed for applications such as auto repair, wood working shops, tire shops, etc. It has a compact footprint of 24"W x 34"D x 58"H.
Catalog No. 50-276-7728 Supplier Grainger Supplier No. 16V891
The Ingersoll Rand Type 30 Reciprocating Air Compressor is a 2 HP electric powered air compressor that provides 7.0 CFM at 90 PSI with a 30 gallon ASME vertical air receiver tank. It has a 115V 1-phase motor with automatic thermal overload protection and pressure switch with gauge. This air compressor is oil-free with a cast iron reciprocating pump and is designed for applications such as auto repair, wood working shops, tire shops, etc. It has a compact footprint of 24"W x 34"D x 58"H.