Learn More
Grainger Electric Air Compressor 7.5 hp 2 Stage
Catalog No. 502767730
Encompass_Preferred
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade air compressor designed for high performance and durability. It features a 7.5 HP electric motor with a two-stage pump that delivers 16.2 CFM at 175 PSI with a maximum pressure of 175 PSI. The 60 gallon vertical ASME air receiver provides extended run times. This air compressor includes an aftercooler and moisture separator to help remove moisture. It has a compact footprint and comes with vibration isolation feet to reduce noise. The pump is oil-lubricated for smooth operation. This air compressor is backed by a 2 year warranty.
Catalog No. 50-276-7730 Supplier Grainger Supplier No. 16V886
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade air compressor designed for high performance and durability. It features a 7.5 HP electric motor with a two-stage pump that delivers 16.2 CFM at 175 PSI with a maximum pressure of 175 PSI. The 60 gallon vertical ASME air receiver provides extended run times. This air compressor includes an aftercooler and moisture separator to help remove moisture. It has a compact footprint and comes with vibration isolation feet to reduce noise. The pump is oil-lubricated for smooth operation. This air compressor is backed by a 2 year warranty.