Learn More
Grainger Electric Air Compressor 7.5 hp 2 Stage
Catalog No. 502766357
Encompass_Preferred
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade air compressor designed for high performance and durability. It features a 7.5 HP electric motor with a two-stage pump that delivers 16.2 CFM at 175 PSI with a maximum pressure of 175 PSI. The 60 gallon vertical ASME air receiver provides extended air storage. It has a cast iron V-twin cylinder pump and is belt driven for quiet operation. This air compressor requires a 230V 1-phase power supply and includes an automatic tank drain to remove moisture. The unit dimensions are 30" L x 34" W x 75" H and it weighs approximately 415 lbs.
Catalog No. 50-276-6357 Supplier Grainger Supplier No. 16V890
The Ingersoll Rand 7.5 HP 60 Gallon Two Stage Electric Air Compressor (Model 2475N7.5) is an industrial-grade air compressor designed for high performance and durability. It features a 7.5 HP electric motor with a two-stage pump that delivers 16.2 CFM at 175 PSI with a maximum pressure of 175 PSI. The 60 gallon vertical ASME air receiver provides extended air storage. It has a cast iron V-twin cylinder pump and is belt driven for quiet operation. This air compressor requires a 230V 1-phase power supply and includes an automatic tank drain to remove moisture. The unit dimensions are 30" L x 34" W x 75" H and it weighs approximately 415 lbs.