Learn More
Daigger & Company ELECTRIC BUNSEN BURNER
SDP
Catalog No. NC9573037 Shop All Fisherbrand Products
Encompass
ELECTRIC BUNSEN BURNER 400 WATT 800-1000C CORROSION RESISTANT METAL HOUSING 7 in.H X 3.5 BASE DIAMETER
Catalog No. NC9573037
ELECTRIC BUNSEN BURNER 400 WATT 800-1000C CORROSION RESISTANT METAL HOUSING 7 in.H X 3.5 BASE DIAMETER