Learn More
Grainger Electric Chain Hoist 10 000 lb 10 ft.
Catalog No. 502749339
Encompass_Preferred
The Dayton Electric Chain Hoist H2 Light is a compact, lightweight hoist for lifting and pulling up to 2 tons. It features a 120V AC motor with thermal overload protection, precision hardened alloy steel load chain, and rugged steel alloy housing. It has a lift speed of 16 feet per minute, lift height up to 20 feet, and a 20-foot lift chain. Operate it with the attached 10-foot cord or hardwire it. Safety features include an automatic brake and overload limiter. UL listed and CSA certified, this hoist is ideal for light-duty lifting applications.
Catalog No. 50-274-9339 Supplier Grainger Supplier No. 14H734
Add to Cart
Add to Cart
Request a Quote
The Dayton Electric Chain Hoist H2 Light is a compact, lightweight hoist for lifting and pulling up to 2 tons. It features a 120V AC motor with thermal overload protection, precision hardened alloy steel load chain, and rugged steel alloy housing. It has a lift speed of 16 feet per minute, lift height up to 20 feet, and a 20-foot lift chain. Operate it with the attached 10-foot cord or hardwire it. Safety features include an automatic brake and overload limiter. UL listed and CSA certified, this hoist is ideal for light-duty lifting applications.