Learn More
Grainger Electric Chain Hoist 10 000 lb 20 ft.
Catalog No. 502752255
Encompass_Preferred
The Harrington Electric Chain Hoist H4 Heavy Duty is a 1/4 ton capacity electric chain hoist with a 4 foot lift. It has a 115V AC single phase motor with a chain speed of 8 feet per minute. The hoist has a steel alloy load chain and hook with latch. It comes with an 8 foot power cord and hand held control. The hoist meets ASME specifications and is UL listed. With a compact, lightweight design, this electric hoist is ideal for light duty lifting applications.
Catalog No. 50-275-2255 Supplier Grainger Supplier No. 14H736
Add to Cart
Add to Cart
Request a Quote
The Harrington Electric Chain Hoist H4 Heavy Duty is a 1/4 ton capacity electric chain hoist with a 4 foot lift. It has a 115V AC single phase motor with a chain speed of 8 feet per minute. The hoist has a steel alloy load chain and hook with latch. It comes with an 8 foot power cord and hand held control. The hoist meets ASME specifications and is UL listed. With a compact, lightweight design, this electric hoist is ideal for light duty lifting applications.