Learn More
Grainger Electric Chain Hoist 10 000 lb 15 ft.
Catalog No. 502748977
Encompass_Preferred
The Dayton Electric Chain Hoist from Grainger is a lifting device that uses an electric motor to lift and lower loads via a chain. It has a lifting capacity of up to 5 tons and comes in multiple voltage options including 115V, 208V, 230V, 460V, and 575V. The hoist can lift at speeds up to 16 feet per minute and has a lift height up to 30 feet. Safety features include a brake, overload protection, limit switch, and top and bottom limit stops. The hoist requires regular lubrication and inspection. Multiple hoist configurations are available including standard, low headroom, and double girder designs.
Catalog No. 50-274-8977 Supplier Grainger Supplier No. 14H735
Add to Cart
Add to Cart
Request a Quote
The Dayton Electric Chain Hoist from Grainger is a lifting device that uses an electric motor to lift and lower loads via a chain. It has a lifting capacity of up to 5 tons and comes in multiple voltage options including 115V, 208V, 230V, 460V, and 575V. The hoist can lift at speeds up to 16 feet per minute and has a lift height up to 30 feet. Safety features include a brake, overload protection, limit switch, and top and bottom limit stops. The hoist requires regular lubrication and inspection. Multiple hoist configurations are available including standard, low headroom, and double girder designs.