Learn More
Grainger Electric Chain Hoist 1000 lb 15 ft.
Catalog No. 502751531
Encompass_Preferred
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor, forged steel load chain, and Weston-style mechanical load brake. This hoist has a lift speed of 8 ft/min and lift height up to 20 ft. It requires 460V AC 3 phase power. The hoist body is made of heavy gauge steel with powder coat paint. It comes with 20 ft of lift chain, hook mounted control pendant, and upper and lower hooks. This is an overhead lifting device designed for rugged industrial lifting applications.
Catalog No. 50-275-1531 Supplier Grainger Supplier No. 14H717
Add to Cart
Add to Cart
Request a Quote
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor, forged steel load chain, and Weston-style mechanical load brake. This hoist has a lift speed of 8 ft/min and lift height up to 20 ft. It requires 460V AC 3 phase power. The hoist body is made of heavy gauge steel with powder coat paint. It comes with 20 ft of lift chain, hook mounted control pendant, and upper and lower hooks. This is an overhead lifting device designed for rugged industrial lifting applications.