Learn More
Grainger Electric Chain Hoist 1000 lb 10 ft.
Catalog No. 502753060
Encompass_Preferred
The Harrington Electric Chain Hoist H4 Heavy Duty is a 1/4 ton capacity electric chain hoist with a 4 foot lift. It has a single phase 115V AC motor with a hoisting speed of 16 feet per minute. The hoist has a forged steel hook with latch and features a Weston-style mechanical load brake. This hoist meets ASME specifications and comes with a 12 month warranty from Harrington. The overall dimensions are 11 x 5 x 11 inches and it weighs 41 lbs. This heavy duty electric hoist is ideal for warehouses, factories, construction sites and other industrial lifting applications.
Catalog No. 50-275-3060 Supplier Grainger Supplier No. 14H713
The Harrington Electric Chain Hoist H4 Heavy Duty is a 1/4 ton capacity electric chain hoist with a 4 foot lift. It has a single phase 115V AC motor with a hoisting speed of 16 feet per minute. The hoist has a forged steel hook with latch and features a Weston-style mechanical load brake. This hoist meets ASME specifications and comes with a 12 month warranty from Harrington. The overall dimensions are 11 x 5 x 11 inches and it weighs 41 lbs. This heavy duty electric hoist is ideal for warehouses, factories, construction sites and other industrial lifting applications.