Learn More
Grainger Electric Chain Hoist 1000 lb 20 ft.
Catalog No. 502751841
Encompass_Preferred
The Grainger Approved Electric Chain Hoist is a 1/4 ton capacity hoist with a lift height of 20 feet. It has a 115V AC single phase motor with a lifting speed of 8 feet per minute. Safety features include an automatic brake, overload protection, and upper and lower limit switches. The hoist comes with a 20 foot lift chain and hook. It has a rugged steel construction and is designed for light duty lifting applications. The hoist requires minimal maintenance and meets ASME standards. With proper installation and operation, this hoist provides a reliable lifting solution.
Catalog No. 50-275-1841 Supplier Grainger Supplier No. 14H715
The Grainger Approved Electric Chain Hoist is a 1/4 ton capacity hoist with a lift height of 20 feet. It has a 115V AC single phase motor with a lifting speed of 8 feet per minute. Safety features include an automatic brake, overload protection, and upper and lower limit switches. The hoist comes with a 20 foot lift chain and hook. It has a rugged steel construction and is designed for light duty lifting applications. The hoist requires minimal maintenance and meets ASME standards. With proper installation and operation, this hoist provides a reliable lifting solution.