Learn More
Grainger Electric Chain Hoist 2000 lb 10 ft.
Catalog No. 502753591
Encompass_Preferred
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor, 30 foot lift, and chain container. This hoist has a lifting speed of 8 feet per minute. It requires 460V 3 phase power and includes top and bottom hooks, load chain, and 10 feet of power cable. The hoist meets ASME specifications and is made in the USA. This is a heavy duty electric hoist suitable for demanding material handling applications.
Catalog No. 50-275-3591 Supplier Grainger Supplier No. 14H722
Add to Cart
Add to Cart
Request a Quote
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor, 30 foot lift, and chain container. This hoist has a lifting speed of 8 feet per minute. It requires 460V 3 phase power and includes top and bottom hooks, load chain, and 10 feet of power cable. The hoist meets ASME specifications and is made in the USA. This is a heavy duty electric hoist suitable for demanding material handling applications.