Learn More
Grainger Electric Chain Hoist 2000 lb 20 ft.
Catalog No. 502753417
Encompass_Preferred
The Harrington Electric Chain Hoist H4 Heavy Duty is a 1/4 ton capacity electric chain hoist with a 4 foot lift. It has a 115V AC single phase motor with a chain speed of 8 feet per minute. The hoist has a steel load chain and hook with latch. It is designed for overhead lifting applications and has an enclosed motor and gearcase. The hoist meets ASME specifications and comes with a 12 month warranty from Harrington.
Catalog No. 50-275-3417 Supplier Grainger Supplier No. 14H724
Add to Cart
Add to Cart
Request a Quote
The Harrington Electric Chain Hoist H4 Heavy Duty is a 1/4 ton capacity electric chain hoist with a 4 foot lift. It has a 115V AC single phase motor with a chain speed of 8 feet per minute. The hoist has a steel load chain and hook with latch. It is designed for overhead lifting applications and has an enclosed motor and gearcase. The hoist meets ASME specifications and comes with a 12 month warranty from Harrington.