Learn More
Grainger Electric Chain Hoist 4000 lb 20 ft.
Catalog No. 502753546
Encompass_Preferred
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor, forged steel hooks with safety latches, Weston-style mechanical load brake, and push button controls. This hoist has dual pawl ratchet mechanisms, hardened steel gears, and meets ASME B30.16 safety standards. The lift speed is 8 ft/min. It requires 460V 3 phase power. This heavy duty electric hoist is ideal for demanding material handling applications.
Catalog No. 50-275-3546 Supplier Grainger Supplier No. 14H727
Add to Cart
Add to Cart
Request a Quote
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor, forged steel hooks with safety latches, Weston-style mechanical load brake, and push button controls. This hoist has dual pawl ratchet mechanisms, hardened steel gears, and meets ASME B30.16 safety standards. The lift speed is 8 ft/min. It requires 460V 3 phase power. This heavy duty electric hoist is ideal for demanding material handling applications.