Learn More
Grainger Electric Chain Hoist 6000 lb 10 ft.
Catalog No. 502752858
Encompass_Preferred
The CM Lodestar H4 Electric Chain Hoist is a compact, lightweight hoist designed for lifting and pulling applications up to 4 tons. It features a three phase motor with thermal overload protection, precision machined gears, and a load brake to hold the load in place. The hoist has a lifting speed of 8/4 fpm and can lift up to 32 feet. It comes with a 20 foot lift of load chain, an upper and lower limit switch, and overload protection. The hoist requires a 3 phase power supply and is made of durable steel construction. It meets ASME specifications.
Catalog No. 50-275-2858 Supplier Grainger Supplier No. 14H731
Add to Cart
Add to Cart
Request a Quote
The CM Lodestar H4 Electric Chain Hoist is a compact, lightweight hoist designed for lifting and pulling applications up to 4 tons. It features a three phase motor with thermal overload protection, precision machined gears, and a load brake to hold the load in place. The hoist has a lifting speed of 8/4 fpm and can lift up to 32 feet. It comes with a 20 foot lift of load chain, an upper and lower limit switch, and overload protection. The hoist requires a 3 phase power supply and is made of durable steel construction. It meets ASME specifications.