Learn More
Grainger Electric Chain Hoist 6000 lb 15 ft.
Catalog No. 502751970
Encompass_Preferred
The Electric Chain Hoist from Grainger is a portable lifting device designed for vertical lifting applications. It features a 1/4 ton capacity, 115V AC operation, 10 ft lift, and 16fpm lifting speed. The hoist has a rugged steel frame and liftwheel brake for controlled lifting. It comes with a 20ft lift chain and hook. The hoist requires minimal maintenance and meets ASME specifications. With portable operation and 1/4 ton capacity, this electric hoist is well-suited for light duty lifting applications.
Catalog No. 50-275-1970 Supplier Grainger Supplier No. 14H732
Add to Cart
Add to Cart
Request a Quote
The Electric Chain Hoist from Grainger is a portable lifting device designed for vertical lifting applications. It features a 1/4 ton capacity, 115V AC operation, 10 ft lift, and 16fpm lifting speed. The hoist has a rugged steel frame and liftwheel brake for controlled lifting. It comes with a 20ft lift chain and hook. The hoist requires minimal maintenance and meets ASME specifications. With portable operation and 1/4 ton capacity, this electric hoist is well-suited for light duty lifting applications.