Learn More
Grainger Electric Chain Hoist 6000 lb 20 ft.
Catalog No. 502752247
Encompass_Preferred
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor and trolley mount for overhead lifting applications. The hoist has a lift speed of 8 ft/min and is made of heavy duty steel construction with a powder coat finish. Safety features include an automatic brake and overload protection. The hoist requires 460V 3 phase power and includes 20 ft of lift chain. This heavy duty electric hoist is ideal for industrial lifting applications requiring up to 4 tons capacity.
Catalog No. 50-275-2247 Supplier Grainger Supplier No. 14H733
Add to Cart
Add to Cart
Request a Quote
The Harrington H4 Heavy Duty Electric Chain Hoist has a lifting capacity of 4 tons (8,000 lbs). It features a 3 phase motor and trolley mount for overhead lifting applications. The hoist has a lift speed of 8 ft/min and is made of heavy duty steel construction with a powder coat finish. Safety features include an automatic brake and overload protection. The hoist requires 460V 3 phase power and includes 20 ft of lift chain. This heavy duty electric hoist is ideal for industrial lifting applications requiring up to 4 tons capacity.