Learn More
MSC CAM Spray Electric Cold Water Pressure Washer, 4 GPM, 2000 PSI, 230V
Catalog No. 502838815
Encompass
The CAM Spray X cart cold water electric power washer features a durable, industrially coated tube steel frame with two 10" tires for excellent portability. It includes an industrial motor with thermal overload protection and a triplex plunger pump with ceramic plungers and stainless steel valves, safeguarded by unloader and secondary pop-off valves. Low-pressure chemical injection is available for applying concentrated liquid detergents.
  • Heavy tube steel frame protected by an industrial coating.
  • Excellent portability with two 10" tires.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump with ceramic plungers and stainless steel valves.
  • Pump protected by unloader valve and secondary pop-off valve.
  • Equipped with low pressure chemical injection, activated by detergent nozzle.
  • Wand and hose rack integrated into frame design.
Catalog No. 50-283-8815 Supplier MSC Supplier No. 63820195
Add to Cart
Add to Cart
Request a Quote
The CAM Spray X cart cold water electric power washer features a durable, industrially coated tube steel frame with two 10" tires for excellent portability. It includes an industrial motor with thermal overload protection and a triplex plunger pump with ceramic plungers and stainless steel valves, safeguarded by unloader and secondary pop-off valves. Low-pressure chemical injection is available for applying concentrated liquid detergents.
  • Heavy tube steel frame protected by an industrial coating.
  • Excellent portability with two 10" tires.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump with ceramic plungers and stainless steel valves.
  • Pump protected by unloader valve and secondary pop-off valve.
  • Equipped with low pressure chemical injection, activated by detergent nozzle.
  • Wand and hose rack integrated into frame design.