Learn More
Grainger Electric Drill 1/2 In 0 to 900 rpm 7.0A
Catalog No. 502749366
Encompass_Preferred
The Grainger 1/2" Electric Drill is a corded, variable speed drill with 0-900 RPM. It has a 7 amp motor providing 265 in-lbs of torque. The drill has a keyed chuck accepting drill bits up to 1/2" diameter. It features an auxiliary handle, lock-on button and overload protection. The overall length is 9-1/2" and it weighs 5 lbs. This electric drill would be suitable for metalworking, woodworking and drilling projects requiring moderate power.
Catalog No. 50-274-9366 Supplier Grainger Supplier No. 14F086
The Grainger 1/2" Electric Drill is a corded, variable speed drill with 0-900 RPM. It has a 7 amp motor providing 265 in-lbs of torque. The drill has a keyed chuck accepting drill bits up to 1/2" diameter. It features an auxiliary handle, lock-on button and overload protection. The overall length is 9-1/2" and it weighs 5 lbs. This electric drill would be suitable for metalworking, woodworking and drilling projects requiring moderate power.