Learn More
Grainger Honeywell Home RedLINK®-Enabled Electrical Heat Equipment Interface Module, 120/240 VAC, 12.5 A, 1 ea
Catalog No. 502771548
Encompass_Preferred
The RedLINK™-Enabled Electrical Heat Equipment Interface Module simplifies the relocation and modernization of electric heating control systems, including baseboard, convector, and fan-forced heaters. This hardwired module integrates with wireless systems to enable precise temperature management and convenient installation without opening walls.
  • Eliminates the need for opening walls during system or control upgrades
  • Supports operating voltages from 100 V to 240 V
  • Compatible with resistive loads up to 12.5 A (1,500 W at 120 V; 3,000 W at 240 V)
  • Features SPST switching action
  • Includes white and light almond antenna covers
  • Operates in temperatures from -20°C to 60°C (-4°F to 140°F)
  • Approved by Underwriters Laboratories Inc. and the Federal Communications Commission
Catalog No. 50-277-1548 Supplier Grainger Supplier No. 14A011
The RedLINK™-Enabled Electrical Heat Equipment Interface Module simplifies the relocation and modernization of electric heating control systems, including baseboard, convector, and fan-forced heaters. This hardwired module integrates with wireless systems to enable precise temperature management and convenient installation without opening walls.
  • Eliminates the need for opening walls during system or control upgrades
  • Supports operating voltages from 100 V to 240 V
  • Compatible with resistive loads up to 12.5 A (1,500 W at 120 V; 3,000 W at 240 V)
  • Features SPST switching action
  • Includes white and light almond antenna covers
  • Operates in temperatures from -20°C to 60°C (-4°F to 140°F)
  • Approved by Underwriters Laboratories Inc. and the Federal Communications Commission