Learn More
Electric Mobility Science Kit
SCI_ED
Catalog No. S07866
Change view
Click to view available options
Electric Mobility Science Kit
Catalog No.
S07866 Electric Mobility Science Kit
1 options
This item is not returnable. View return policy
Catalog No. S07866 Supplier Horizon Fuel Cell Technologies Supplier No. FCJJ30
This item is not returnable. View return policy
Includes
  • Car chassis with adapter base
  • Silicon tubing
  • Connector wires
  • Hand crank
  • Super capacitor
  • Hydrostik™ PRO cartridge with regulator (empty)
  • Salt water fuel cell
  • Micro-PEM fuel cell (H2/air)
  • Micro-PEM fuel cell (H2/02)
  • Water and O2/H2 storage
  • Battery bank
  • Solar cell
Requires
Hydrostik PRO cartridge is shipped empty. Hydrofill™ PRO S07867 (sold separately) or access to hydrogen is required. Additional Hydrostik PRO cartridges available separately.
Six different models can be built with one chassis.
  • Examines the strengths and weaknesses of each model
  • Compatible with all other Horizon kits, for measurement, data logging and more
  • Features fixed steering function at front of the vehicle
Specifications
For Use With (Equipment) Other horizon kits
Includes Car chassis with adapter base, Silicon tubing, connector wires, Hand crank, Super capacitor, Hydrostik PRO cartridge, with regulator (empty), salt water fuel cell, Micro-PEM fuel cell, Micro-PEM fuel cell, water and O2/H2 storage, battery bank, solar cell
Product Type Electric Mobility Science Kit