Learn More
Bellco Glass, Inc. Electric Pipet Controller
SDP
Catalog No. 502330851
Encompass_Preferred
Lightweight (6.3oz/170g) and designed to fit comfortably in either hand. Long-life rechargeable lithium battery provides up to 8 hours of cordless use on a 2 hour charge.\n\nSpecifications:\n
    \n
  • Adjustable dial for speed setting
  • \n
  • Low pressure fingertip control buttons
  • \n
  • Easy fit adapter accepts pipet sizes from 1 to 100mL
  • \n
Catalog No. 50-233-0851 Supplier Bellco Glass, Inc. Supplier No. 230200000
Lightweight (6.3oz/170g) and designed to fit comfortably in either hand. Long-life rechargeable lithium battery provides up to 8 hours of cordless use on a 2 hour charge.\n\nSpecifications:\n
    \n
  • Adjustable dial for speed setting
  • \n
  • Low pressure fingertip control buttons
  • \n
  • Easy fit adapter accepts pipet sizes from 1 to 100mL
  • \n