Learn More
Protexer Corporation Electric Shoe Cover Refill - PE 2,100 pairs/Case
Supplier:  Protexer Corporation EB2100PE
Encompass_Preferred
For use with Electric BootieButler Dispensers (EMLG) only. Impervious shoe cover Constructed of Polyethylene material. One size fits most (up to men’s size 15). Approved for Class 10,000/ISO-7 Cleanrooms.
Catalog No. NC2228099