Learn More
Arrow Engineering Direct Drive Overhead Stirrer, Electric Model JR4000, 120V 60HZ
Catalog No. NC2948072
Encompass_Preferred
The Arrow Mixing Products Model JR4000 Electric Stirrer is the perfect choice for light-duty mixing applications. Designed with precision and efficiency in mind, this versatile overhead mixer is ideal for laboratories, small-scale production, and various other settings where reliable mixing performance is essential. This 1/25HP mixer can run at speeds from 80-4,000 RPM's. The Model JR4000 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade(modfied pitch) propeller & aluminum coupling.
Catalog No. NC2948072 Supplier Arrow Engineering Supplier No. JR4000
The Arrow Mixing Products Model JR4000 Electric Stirrer is the perfect choice for light-duty mixing applications. Designed with precision and efficiency in mind, this versatile overhead mixer is ideal for laboratories, small-scale production, and various other settings where reliable mixing performance is essential. This 1/25HP mixer can run at speeds from 80-4,000 RPM's. The Model JR4000 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade(modfied pitch) propeller & aluminum coupling.