Learn More
Grainger Electric Tankless Water Heater 208V
Catalog No. 502751585
Encompass_Preferred
The Eemax Electric Tankless Water Heater is a point-of-use electric water heater that provides hot water instantly and on demand. It has a compact, space-saving design and is available in power ratings from 2.5kW to 27kW. The unit has an all copper heating system, adjustable digital temperature control, and self-modulating technology to maintain the set temperature. Safety features include overheat protection, no flow shutdown, and freeze protection. The water heater can be wall mounted or installed under a sink and has a stainless steel enclosure. It's ideal for residential and commercial applications where hot water is needed at a single point of use.
Catalog No. 50-275-1585 Supplier Grainger Supplier No. 14U404
The Eemax Electric Tankless Water Heater is a point-of-use electric water heater that provides hot water instantly and on demand. It has a compact, space-saving design and is available in power ratings from 2.5kW to 27kW. The unit has an all copper heating system, adjustable digital temperature control, and self-modulating technology to maintain the set temperature. Safety features include overheat protection, no flow shutdown, and freeze protection. The water heater can be wall mounted or installed under a sink and has a stainless steel enclosure. It's ideal for residential and commercial applications where hot water is needed at a single point of use.