Learn More
Grainger Electric Tankless Water Heater 240V
Catalog No. 502750395
Encompass_Preferred
The Grainger Approved Electric Tankless Water Heater is a point-of-use electric water heater that provides hot water on demand without a storage tank. It has a stainless steel construction and is available in 4.5 kW or 6 kW power levels. The 4.5 kW model provides up to 1.5 GPM of hot water at a 45 degree F temperature rise. The 6 kW model provides up to 2 GPM. It has an activation flow rate of 0.3 GPM and is designed for indoor installation. The water heater has adjustable temperature settings from 80 to 140 degrees Fahrenheit. It measures 13.75 x 11 x 3.5 inches and weighs 11 pounds. The tankless design provides an endless supply of hot water and helps conserve energy.
Catalog No. 50-275-0395 Supplier Grainger Supplier No. 14U406
The Grainger Approved Electric Tankless Water Heater is a point-of-use electric water heater that provides hot water on demand without a storage tank. It has a stainless steel construction and is available in 4.5 kW or 6 kW power levels. The 4.5 kW model provides up to 1.5 GPM of hot water at a 45 degree F temperature rise. The 6 kW model provides up to 2 GPM. It has an activation flow rate of 0.3 GPM and is designed for indoor installation. The water heater has adjustable temperature settings from 80 to 140 degrees Fahrenheit. It measures 13.75 x 11 x 3.5 inches and weighs 11 pounds. The tankless design provides an endless supply of hot water and helps conserve energy.