Learn More
Grainger Buyers Products Electric Side Mount Tarp System, Aluminum, 5-Spring, 14 ft. to 23 ft. Dump Bodies, 1 ea.
Catalog No. 502785577
Encompass_Preferred
This electric side mount tarp system is designed to secure payloads in dump bodies ranging from 14 ft to 23 ft. Engineered for durability, the system features a 5-spring aluminum construction that keeps materials contained and protected from the elements during transport. The kit includes all necessary installation hardware and wire connectors, utilizing a high-speed, bi-directional 600 W motor with a 90:1 gear ratio for reliable operation.
  • Designed for dump body lengths of 14 ft to 23 ft
  • Features a 90:1 gear ratio and 600 W, bi-directional motor
  • Includes a wind deflector
  • Comes with mounting hardware and wire connectors
  • Constructed from durable aluminum
Catalog No. 50-278-5577 Supplier Grainger Supplier No. 19A791
This electric side mount tarp system is designed to secure payloads in dump bodies ranging from 14 ft to 23 ft. Engineered for durability, the system features a 5-spring aluminum construction that keeps materials contained and protected from the elements during transport. The kit includes all necessary installation hardware and wire connectors, utilizing a high-speed, bi-directional 600 W motor with a 90:1 gear ratio for reliable operation.
  • Designed for dump body lengths of 14 ft to 23 ft
  • Features a 90:1 gear ratio and 600 W, bi-directional motor
  • Includes a wind deflector
  • Comes with mounting hardware and wire connectors
  • Constructed from durable aluminum