Learn More
Grainger Electric Winch 4-4/5HP 12VDC
Catalog No. 502766229
Encompass_Preferred
The WARN 16Y216 Electric Winch is a 12V DC electric winch with a pulling capacity of 2,500 lbs. It has a 50 ft steel wire rope and a 3-stage planetary gear system for smooth, efficient pulling. The winch is powered by a permanent magnet motor that draws 45 amps at stall load. It features a sliding clutch for free spooling, an automatic load-holding brake, and an ergonomic clutch handle. The winch is designed for ATV/UTV use and includes a roller fairlead, mounting channel and hardware. It weighs 22 lbs. Overall, this heavy-duty electric winch provides powerful pulling capacity for ATV/UTV recovery and utility work.
Catalog No. 50-276-6229 Supplier Grainger Supplier No. 16Y216
The WARN 16Y216 Electric Winch is a 12V DC electric winch with a pulling capacity of 2,500 lbs. It has a 50 ft steel wire rope and a 3-stage planetary gear system for smooth, efficient pulling. The winch is powered by a permanent magnet motor that draws 45 amps at stall load. It features a sliding clutch for free spooling, an automatic load-holding brake, and an ergonomic clutch handle. The winch is designed for ATV/UTV use and includes a roller fairlead, mounting channel and hardware. It weighs 22 lbs. Overall, this heavy-duty electric winch provides powerful pulling capacity for ATV/UTV recovery and utility work.