Learn More
Heidolph™ Electrical Cylinder for Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900956 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-956 Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-956 Supplier Heidolph™ Supplier No. 036280120
Add to Cart
Add to Cart
Request a Quote
Electrical Cylinder lowers and raises the heating bath.
Included with Hei-VAP Industrial Rotary Evaporator
Specifications
Type Accessory
For Use With (Equipment) Hei-VAP Industrial Rotary Evaporator
Voltage 24 VDC
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov