Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
| Product Type | Electrical Risk Lockouts |
| Includes | (1) single-pole circuit breaker; (2) multi-pole circuit breaker; (2) no-hole circuit breaker; (1) multi-pole circuit breaker; (2) wall switch lockouts |
| Quantity | 1 Kit |