Learn More
Uline Electrical Tape, Purple
Catalog No. NC2383841
Encompass_Preferred
Electrical tape for splicing and insulation, Rubber adhesive with soft polyvinyl chloride film backing, 1 1/4 core, Stretches up to 200%, Apply in temperatures as low as 32F
Catalog No. NC2383841 Supplier Uline Supplier No. S13516
Electrical tape for splicing and insulation, Rubber adhesive with soft polyvinyl chloride film backing, 1 1/4 core, Stretches up to 200%, Apply in temperatures as low as 32F