Learn More
MSC ELECTRICAL TAPE ROLL
Catalog No. 5000769892
Encompass
This electrical tape features a durable vinyl adhesive, designed to maintain vivid color even when exposed to abrasion and solvents. Its conformable backing makes it ideal for use on curved and irregular surfaces, while the rubber adhesive ensures good instant adhesion on many surfaces.
  • Maintains vivid color even when exposed to abrasion and solvents
  • Available in a wide variety of colors for diverse applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas
Catalog No. 50-007-69892 Supplier MSC Supplier No. 10610541
This electrical tape features a durable vinyl adhesive, designed to maintain vivid color even when exposed to abrasion and solvents. Its conformable backing makes it ideal for use on curved and irregular surfaces, while the rubber adhesive ensures good instant adhesion on many surfaces.
  • Maintains vivid color even when exposed to abrasion and solvents
  • Available in a wide variety of colors for diverse applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas