Learn More
Grainger IPA Electrical Terminal Maintenance Kit, Diamond-Coated Cleaners, Holster, 1 ea.
Catalog No. 502783048
Encompass_Preferred
The HD Fleet Terminal Chaser™ Master Set is a comprehensive maintenance solution designed for cleaning and repairing electrical connectors on heavy-duty vehicles, trucks, trailers, and industrial equipment. This patented tool set effectively removes corrosion from male and female terminals to ensure proper conductivity and prevent intermittent power failures, ultimately extending the life of electrical harnesses.
  • Cleans a wide range of connector types including Deutsch, Cannon, Weather Pack, 7-way round, ABS sensor plugs, and bullet connectors
  • Includes two sizes of flat male and female terminal cleaners, two gold 7-way round pin socket harness cleaners, one black ABS sensor/power connector cleaner, ten micro diamond round files, one trailer light bullet connector cleaner, and DeoxIT® contact cleaner
  • Features diamond-coated tools for precise, effective restoration of electrical terminals from .03 i
Catalog No. 50-278-3048 Supplier Grainger Supplier No. 19RE94
The HD Fleet Terminal Chaser™ Master Set is a comprehensive maintenance solution designed for cleaning and repairing electrical connectors on heavy-duty vehicles, trucks, trailers, and industrial equipment. This patented tool set effectively removes corrosion from male and female terminals to ensure proper conductivity and prevent intermittent power failures, ultimately extending the life of electrical harnesses.
  • Cleans a wide range of connector types including Deutsch, Cannon, Weather Pack, 7-way round, ABS sensor plugs, and bullet connectors
  • Includes two sizes of flat male and female terminal cleaners, two gold 7-way round pin socket harness cleaners, one black ABS sensor/power connector cleaner, ten micro diamond round files, one trailer light bullet connector cleaner, and DeoxIT® contact cleaner
  • Features diamond-coated tools for precise, effective restoration of electrical terminals from .03 i