Learn More
MSC Gleason Reel Wire Reel Cart, Aluminum Frame, 300 lb. Capacity, 3 Slots, 1 ea.
Catalog No. 5000513294
Encompass
This wire reel utility cart is designed for efficient organization and transport of wire spools in industrial environments. Featuring a robust extruded aluminum frame, the cart provides a reliable solution for heavy-duty applications with a maximum load capacity of 300 lbs.
  • Strong extruded aluminum frame
  • Three wire spool slots with 27 in. of clear space between brackets
  • Chip-resistant baked polyester finish
  • Includes four heavy-duty casters, with two fixed and two locking swivel wheels
  • Equipped with a bin bar for storage of wire nuts and fasteners
  • Formed steel brackets provide high rigidity
  • Requires assembly
Catalog No. 50-005-13294 Supplier MSC Supplier No. 00496091
This wire reel utility cart is designed for efficient organization and transport of wire spools in industrial environments. Featuring a robust extruded aluminum frame, the cart provides a reliable solution for heavy-duty applications with a maximum load capacity of 300 lbs.
  • Strong extruded aluminum frame
  • Three wire spool slots with 27 in. of clear space between brackets
  • Chip-resistant baked polyester finish
  • Includes four heavy-duty casters, with two fixed and two locking swivel wheels
  • Equipped with a bin bar for storage of wire nuts and fasteners
  • Formed steel brackets provide high rigidity
  • Requires assembly