Learn More
Julabo USA Inc Bath Accessories, Julabo, Electronic module with analog connections
SDP
Catalog No. 507534129
Encompass_Preferred
Bath Accessories, Julabo, Electronic module with one input and two outputs for external programming or flow sensor or temperature recorder (current/voltage), scalable as well as Standby input and Alarm output. Available as option or to retrofit for circulators of the HighTech series. For use with CF31, CF41, F25-HE, F25-HL, F32-HL, F33-HL, F34-HE, F95-SL, FP35-HL, FP50-HE, FP50-HL, FP52-SL, FP52-SL, FP52-SL-150C, FP55-SL, FP55-SL, FP55-SL-150C, FP89-HL, FP90-SL, FP90-SL, FP90-SL-150C, FPW52-SL, FPW52-SL, FPW52-SL-150C, FPW55-SL, FPW55-SL, FPW55-SL-150C, FPW90-SL, FPW90-SL, FPW90-SL-150C, FPW91-SL, FPW91-SL, FW95-SL, HE-4, HL-4, SE-12, SE-26, SE-6, SE-Z, SL-12, SL-26, SL-6
Catalog No. 50-753-4129 Supplier Julabo USA Inc Supplier No. 8900100
Bath Accessories, Julabo, Electronic module with one input and two outputs for external programming or flow sensor or temperature recorder (current/voltage), scalable as well as Standby input and Alarm output. Available as option or to retrofit for circulators of the HighTech series. For use with CF31, CF41, F25-HE, F25-HL, F32-HL, F33-HL, F34-HE, F95-SL, FP35-HL, FP50-HE, FP50-HL, FP52-SL, FP52-SL, FP52-SL-150C, FP55-SL, FP55-SL, FP55-SL-150C, FP89-HL, FP90-SL, FP90-SL, FP90-SL-150C, FPW52-SL, FPW52-SL, FPW52-SL-150C, FPW55-SL, FPW55-SL, FPW55-SL-150C, FPW90-SL, FPW90-SL, FPW90-SL-150C, FPW91-SL, FPW91-SL, FW95-SL, HE-4, HL-4, SE-12, SE-26, SE-6, SE-Z, SL-12, SL-26, SL-6