Learn More
Waters Corp Electrochemical Detector, PQ Mobile Phase, High Sensitivity, Wide Linear Range
Catalog No. 50792729 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Electrochemical Detector PQ Mobile Phase is an electrochemical detector for high-performance liquid chromatography (HPLC) systems. It converts electrochemical responses of analytes into precise signals, amplified and displayed on the connected HPLC system.
  • Detects broad spectrum analytes: organic acids, bases, and neutral compounds.
  • High sensitivity, <10 nanograms per milliliter detection limit for specific analytes.
  • Applicable across biopharmaceutical, pharmaceutical, environmental, and food analysis.
  • Wide linear range, spanning up to 5 orders of magnitude.
  • Rapid response time, <10 milliseconds, for swift data acquisition.
  • Effortless installation and operation.
  • Consistent performance and quality.
Catalog No. 50-792-729 Supplier Waters Corp Supplier No. 700002287
The Electrochemical Detector PQ Mobile Phase is an electrochemical detector for high-performance liquid chromatography (HPLC) systems. It converts electrochemical responses of analytes into precise signals, amplified and displayed on the connected HPLC system.
  • Detects broad spectrum analytes: organic acids, bases, and neutral compounds.
  • High sensitivity, <10 nanograms per milliliter detection limit for specific analytes.
  • Applicable across biopharmaceutical, pharmaceutical, environmental, and food analysis.
  • Wide linear range, spanning up to 5 orders of magnitude.
  • Rapid response time, <10 milliseconds, for swift data acquisition.
  • Effortless installation and operation.
  • Consistent performance and quality.