Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrochemical Standards, 0.5M DIPA-HCL-ISA, Mettler Toledo™

Catalog No. 01912568 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-912-568 250 mL
01-912-567 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-912-568 Supplier METTLER TOLEDO Supplier No. 30066674
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Sodium analyzer

The new Sodium Analyzer determines the sodium content in food products simply and quickly. Reduce sample preparation time and effort by using safe and cheap chemicals while specifically and exclusively measuring sodium ions.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 0.5M DIPA-HCL-ISA
Packaging Cardboard
CAS 108-18-9, 819-79-4, 7732-18-5
Molecular Formula C6H16ClNO2
Color Colorless
pH 11.4
Boiling Point Approx. 100°C
Solution Type Calibration Solution
Industry Type Verification
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.