Learn More
LGC GENOMICS LLC TransforMax™ EPI300™ Electrocompetent E. coli, High Transformation Efficiency >1 x 10^10 cfu/µg, Inducible Copy Number Control, 10 x 100 µL
Catalog No. NC2893052
Encompass
TransforMax™ EPI300™ Electrocompetent *E. coli* are designed for efficient cloning of large DNA inserts, achieving transformation efficiencies greater than 1 x 10^10 cfu/µg pUC19 DNA. They support inducible copy number control and are excellent for Gibson Assembly reactions. The cells are optimized for high DNA yields and enhanced stability of large constructs, enabling reliable cloning with convenient blue/white screening.
Catalog No. NC2893052 Supplier LGC GENOMICS LLC Supplier No. EC300110
TransforMax™ EPI300™ Electrocompetent *E. coli* are designed for efficient cloning of large DNA inserts, achieving transformation efficiencies greater than 1 x 10^10 cfu/µg pUC19 DNA. They support inducible copy number control and are excellent for Gibson Assembly reactions. The cells are optimized for high DNA yields and enhanced stability of large constructs, enabling reliable cloning with convenient blue/white screening.