Learn More
METTLER TOLEDO Puncture Electrode LoT406
Catalog No. NC9808209 Shop All METTLER TOLEDO Products
Encompass
LoT406-M6-DXK is a puncture electrode with a polymer shaft and XEROLYT PLUS polymer electrolyte specially designed for pH measurement in meat, sausage and cheese.
Catalog No. NC9808209 Supplier METTLER TOLEDO Supplier No. 59903311
LoT406-M6-DXK is a puncture electrode with a polymer shaft and XEROLYT PLUS polymer electrolyte specially designed for pH measurement in meat, sausage and cheese.