Learn More
World Precision Instrument Electrode Adapter for EVOM2
SDP
Catalog No. 502521465
Encompass
Do you want to connect a different electrode to your EVOM2? This adaptor fits into the EVOM2 RJ-11 plug and provides a 2mm pin port for each of the four RJ-11 leads.
Catalog No. 50-252-1465 Supplier World Precision Instrument Supplier No. 3993
Do you want to connect a different electrode to your EVOM2? This adaptor fits into the EVOM2 RJ-11 plug and provides a 2mm pin port for each of the four RJ-11 leads.