Learn More
Cole-Parmer Oakton Ion Selective Electrode (Electrode type: Ammonia)
Catalog No. 501961836 Shop All Cole-Parmer Products
Encompass_Preferred
A wide variety of ion selective electrodes are availabe to suit a wide array of applications. Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds. Epoxy-body ISEs are either single- or double-junction, and the glass-body type is double junction.
Catalog No. 50-196-1836 Supplier Cole-Parmer Supplier No. 3580200
A wide variety of ion selective electrodes are availabe to suit a wide array of applications. Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds. Epoxy-body ISEs are either single- or double-junction, and the glass-body type is double junction.